Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: loqs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038814.3 | NP_001033903.1 | 4 - 5 | 1457 | 122 | |
NM_135802.4 | NP_609646.1 | 4 - 5 | 1327 | 122 | ||
GenBank mRNA | AY071070.1 | - | 4 - 5 | 1324 | 122 | |
DQ118034.1 | - | 4 - 5 | 1331 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Drosophila melanogaster |