Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp28a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_135850.3 | NP_609694.1 | 4 - 5 | 1142 | 64 | |
GenBank mRNA | AY051533.1 | - | 4 - 5 | 1118 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |