Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZnT35C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_135897.5 | NP_609741.3 | 5 - 6 | 743 | 54 | |
NM_165105.3 | NP_723901.1 | 4 - 5 | 815 | 54 | ||
GenBank mRNA | AY071460.1 | - | 5 - 6 | 740 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |