Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ChLD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_135962.3 | NP_609806.1 | 2 - 3 | 1073 | 74 | |
GenBank mRNA | AY118355.1 | - | 2 - 3 | 1071 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |