Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bur | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299191.1 | NP_001286120.1 | 3 - 4 | 766 | 92 | |
NM_138759.4 | NP_620114.3 | 3 - 4 | 766 | 92 | ||
GenBank mRNA | AY118572.1 | - | 3 - 4 | 743 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |