Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rgn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259937.2 | NP_001246866.1 | 3 - 4 | 1090 | 77 | |
NM_001275226.1 | NP_001262155.1 | 4 - 5 | 1139 | 77 | ||
NM_001275227.1 | NP_001262156.1 | 3 - 4 | 1090 | 77 | ||
NM_141055.4 | NP_649312.1 | 4 - 5 | 1139 | 77 | ||
GenBank mRNA | AF145671.1 | - | 4 - 5 | 1116 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |