Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sod3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038850.2 | NP_001033939.2 | 2 - 3 | 173 | 69 | |
NM_001043071.2 | NP_001036536.1 | 2 - 3 | 173 | 69 | ||
NM_001316415.1 | NP_001303344.1 | 2 - 3 | 173 | 69 | ||
NM_136838.3 | NP_610682.2 | 3 - 4 | 317 | 69 | ||
NM_165829.3 | NP_725046.2 | 2 - 3 | 173 | 69 | ||
GenBank mRNA | BT003225.1 | - | 3 - 4 | 301 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |