Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Drip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273961.1 | NP_001260890.1 | 2 - 3 | 545 | 76 | |
NM_001273962.1 | NP_001260891.1 | 2 - 3 | 319 | 76 | ||
NM_001273963.1 | NP_001260892.1 | 2 - 3 | 211 | 76 | ||
NM_001273964.1 | NP_001260893.1 | 2 - 3 | 776 | 76 | ||
NM_001299387.1 | NP_001286316.1 | 2 - 3 | 776 | 76 | ||
NM_078973.3 | NP_523697.1 | 3 - 4 | 475 | 76 | ||
NM_165833.3 | NP_725051.2 | 2 - 3 | 776 | 76 | ||
GenBank mRNA | BT023883.1 | - | 2 - 3 | 773 | 76 | |
BT126290.1 | - | 2 - 3 | 526 | 76 | ||
JN791442.1 | - | 2 - 3 | 199 | 76 | ||
JN791443.1 | - | 2 - 3 | 199 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Drosophila melanogaster |