Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: wal | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057627.4 | NP_476975.1 | 2 - 3 | 118 | 63 | |
NM_165847.2 | NP_725073.1 | 2 - 3 | 118 | 63 | ||
GenBank mRNA | AF132155.1 | - | 2 - 3 | 101 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |