Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: muskelin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273992.1 | NP_001260921.1 | 2 - 3 | 532 | 69 | |
NM_136957.3 | NP_610801.1 | 2 - 3 | 532 | 69 | ||
NM_165928.1 | NP_725200.1 | 3 - 4 | 566 | 69 | ||
GenBank mRNA | BT150328.1 | - | 3 - 4 | 565 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |