Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001201987.2 | NP_001188916.1 | 4 - 5 | 1070 | 76 | |
NM_058084.5 | NP_477432.3 | 4 - 5 | 1447 | 76 | ||
NM_165947.2 | NP_725226.2 | 4 - 5 | 1296 | 76 | ||
NM_165948.2 | NP_725227.2 | 4 - 5 | 1038 | 76 | ||
GenBank mRNA | AY071059.1 | - | 4 - 5 | 1444 | 76 | |
BT132865.1 | - | 4 - 5 | 1038 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |