Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TppII | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299447.1 | NP_001286376.1 | 5 - 6 | 1808 | 66 | |
NM_001299448.1 | NP_001286377.1 | 5 - 6 | 1783 | 66 | ||
NM_057899.3 | NP_477247.1 | 4 - 5 | 1720 | 66 | ||
NM_165966.2 | NP_725252.1 | 5 - 6 | 2074 | 66 | ||
GenBank mRNA | BT003260.1 | - | 4 - 5 | 1712 | 66 | |
BT044288.1 | - | 5 - 6 | 2025 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |