Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274020.1 | NP_001260949.1 | 5 - 6 | 747 | 64 | |
NM_057691.4 | NP_477039.1 | 5 - 6 | 1360 | 64 | ||
NM_165974.3 | NP_725267.1 | 4 - 5 | 825 | 64 | ||
GenBank mRNA | AY069526.1 | - | 5 - 6 | 1309 | 64 | |
BT056266.1 | - | 4 - 5 | 823 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |