Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dh31-R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274021.2 | NP_001260950.1 | 3 - 4 | 995 | 58 | |
NM_001274022.2 | NP_001260951.1 | 3 - 4 | 995 | 58 | ||
NM_165979.3 | NP_725278.1 | 3 - 4 | 995 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor Drosophila melanogaster |