Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scaf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014589.4 | NP_001014589.1 | 12 - 13 | 2902 | 59 | |
NM_001038939.2 | NP_001034028.2 | 11 - 12 | 2829 | 59 | ||
GenBank mRNA | BT031129.1 | - | 12 - 13 | 2868 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |