Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cad74A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259885.1 | NP_001246814.1 | 6 - 7 | 4504 | 70 | |
NM_140716.2 | NP_648973.1 | 6 - 7 | 4218 | 70 | ||
NM_168724.2 | NP_730261.1 | 6 - 7 | 4233 | 70 | ||
GenBank mRNA | BT150474.1 | - | 6 - 7 | 4232 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |