Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eip74EF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014590.2 | NP_001014590.1 | 2 - 3 | 1897 | 102 | |
NM_001259887.2 | NP_001246816.1 | 6 - 7 | 3156 | 102 | ||
NM_168739.3 | NP_730286.3 | 6 - 7 | 3156 | 102 | ||
NM_168740.3 | NP_730287.1 | 6 - 7 | 3156 | 102 | ||
NM_168741.2 | NP_730288.1 | 2 - 3 | 2203 | 102 | ||
GenBank mRNA | BT133150.1 | - | 2 - 3 | 1488 | 102 | |
M37082.1 | - | 6 - 7 | 3623 | 102 | ||
M37083.1 | - | 2 - 3 | 2459 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |