Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144516.2 | NP_001137988.1 | 6 - 7 | 1077 | 84 | |
NM_001275181.1 | NP_001262110.1 | 6 - 7 | 1121 | 84 | ||
NM_001275182.1 | NP_001262111.1 | 6 - 7 | 1091 | 84 | ||
NM_079460.2 | NP_524184.1 | 6 - 7 | 1248 | 84 | ||
NM_168856.3 | NP_730535.1 | 6 - 7 | 1091 | 84 | ||
GenBank mRNA | AF017024.1 | - | 6 - 7 | 1077 | 84 | |
AY061316.1 | - | 6 - 7 | 1248 | 84 | ||
U77934.1 | - | 5 - 6 | 1021 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |