Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pitslre | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259922.2 | NP_001246851.1 | 3 - 4 | 3723 | 70 | |
NM_001259923.2 | NP_001246852.1 | 3 - 4 | 2952 | 70 | ||
NM_001275193.1 | NP_001262122.1 | 2 - 3 | 3271 | 70 | ||
NM_140994.4 | NP_649251.2 | 2 - 3 | 2593 | 70 | ||
NM_168869.3 | NP_730563.1 | 3 - 4 | 2534 | 70 | ||
GenBank mRNA | AY128425.1 | - | 1 - 2 | 3046 | 70 | |
BT016092.1 | - | 2 - 3 | 2584 | 70 | ||
BT120362.1 | - | 3 - 4 | 3685 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |