Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: siz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001043153.2 | NP_001036618.1 | 5 - 6 | 3432 | 58 | |
NM_001170016.1 | NP_001163487.1 | 5 - 6 | 3709 | 58 | ||
NM_168881.3 | NP_730594.1 | 5 - 6 | 3908 | 58 | ||
NM_206407.2 | NP_996129.1 | 6 - 7 | 4092 | 58 | ||
GenBank mRNA | AY375487.1 | - | 5 - 6 | 3904 | 58 | |
AY375488.1 | - | 5 - 6 | 3852 | 58 | ||
AY375489.1 | - | 5 - 6 | 3432 | 58 | ||
BT150471.1 | - | 5 - 6 | 3904 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |