Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sick | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273674.1 | NP_001260603.1 | 4 - 5 | 1244 | 70 | |
NM_001273677.1 | NP_001260606.1 | 3 - 4 | 2263 | 70 | ||
NM_001273678.1 | NP_001260607.1 | 3 - 4 | 2949 | 70 | ||
NM_001273680.2 | NP_001260609.1 | 4 - 5 | 1149 | 70 | ||
NM_001273681.1 | NP_001260610.1 | 2 - 3 | 702 | 70 | ||
NM_206007.5 | NP_995729.2 | 4 - 5 | 1149 | 70 | ||
NM_206008.4 | NP_995730.2 | 4 - 5 | 3116 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |