Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273683.1 | NP_001260612.1 | 2 - 3 | 503 | 67 | |
NM_001273684.1 | NP_001260613.1 | 1 - 2 | 860 | 67 | ||
NM_136169.4 | NP_610013.1 | 2 - 3 | 477 | 67 | ||
NM_206010.3 | NP_995732.1 | 2 - 3 | 481 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |