Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vari | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273716.1 | NP_001260645.1 | 8 - 9 | 1621 | 73 | |
NM_165342.4 | NP_724288.3 | 9 - 10 | 2022 | 73 | ||
NM_165343.3 | NP_724289.2 | 8 - 9 | 1562 | 73 | ||
NM_206011.3 | NP_995733.2 | 9 - 10 | 1959 | 73 | ||
GenBank mRNA | BT056231.1 | - | 9 - 10 | 1958 | 73 | |
BT132965.1 | - | 9 - 10 | 2020 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |