Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: jing | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103729.2 | NP_001097199.1 | 4 - 5 | 4225 | 101 | |
NM_001103730.2 | NP_001097200.1 | 4 - 5 | 5496 | 101 | ||
NM_136371.3 | NP_610215.3 | 4 - 5 | 4225 | 101 | ||
NM_206027.3 | NP_995749.2 | 4 - 5 | 3916 | 101 | ||
GenBank mRNA | AF285778.1 | - | 3 - 4 | 3689 | 101 | |
BT025201.1 | - | 4 - 5 | 3896 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |