Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273801.1 | NP_001260730.1 | 3 - 4 | 655 | 52 | |
NM_165479.2 | NP_724481.1 | 3 - 4 | 655 | 52 | ||
NM_165480.2 | NP_724482.1 | 2 - 3 | 782 | 52 | ||
NM_206028.2 | NP_995750.1 | 3 - 4 | 662 | 52 | ||
GenBank mRNA | AY069387.1 | - | 2 - 3 | 756 | 52 | |
BT088961.1 | - | 3 - 4 | 663 | 52 | ||
M37787.1 | - | 2 - 3 | 769 | 52 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |