Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vimar | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057957.5 | NP_477305.1 | 2 - 3 | 1209 | 66 | |
NM_206045.2 | NP_995767.1 | 2 - 3 | 1212 | 66 | ||
GenBank mRNA | BT044558.1 | - | 2 - 3 | 1209 | 66 | |
BT125966.1 | - | 2 - 3 | 1180 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |