Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldh-III | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273843.1 | NP_001260772.1 | 2 - 3 | 206 | 89 | |
NM_001273844.1 | NP_001260773.1 | 2 - 3 | 333 | 89 | ||
NM_001273846.1 | NP_001260775.1 | 2 - 3 | 206 | 89 | ||
NM_136441.4 | NP_610285.5 | 4 - 5 | 672 | 89 | ||
NM_165526.3 | NP_724560.1 | 1 - 2 | 405 | 89 | ||
NM_165527.3 | NP_724561.2 | 2 - 3 | 244 | 89 | ||
NM_165528.3 | NP_724562.2 | 4 - 5 | 1193 | 89 | ||
NM_165529.3 | NP_724563.2 | 2 - 3 | 152 | 89 | ||
NM_165530.3 | NP_724564.2 | 2 - 3 | 333 | 89 | ||
NM_165531.3 | NP_724565.3 | 4 - 5 | 548 | 89 | ||
NM_165532.3 | NP_724566.3 | 4 - 5 | 452 | 89 | ||
NM_206050.3 | NP_995772.1 | 2 - 3 | 206 | 89 | ||
GenBank mRNA | BT003248.1 | - | 1 - 2 | 414 | 89 | |
BT003252.1 | - | 2 - 3 | 301 | 89 | ||
BT044325.1 | - | 2 - 3 | 205 | 89 | ||
BT126071.1 | - | 4 - 5 | 699 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |