Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lig | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014503.2 | NP_001014503.1 | 11 - 12 | 3818 | 59 | |
NM_001169610.2 | NP_001163081.1 | 12 - 13 | 3811 | 59 | ||
NM_001169611.1 | NP_001163082.1 | 12 - 13 | 3835 | 59 | ||
NM_001169612.2 | NP_001163083.1 | 13 - 14 | 3877 | 59 | ||
NM_001259271.2 | NP_001246200.1 | 12 - 13 | 3715 | 59 | ||
NM_001273865.1 | NP_001260794.1 | 11 - 12 | 3879 | 59 | ||
NM_001273866.2 | NP_001260795.1 | 10 - 11 | 3598 | 59 | ||
NM_001316409.1 | NP_001303338.1 | 10 - 11 | 3598 | 59 | ||
NM_136504.5 | NP_610348.3 | 12 - 13 | 3811 | 59 | ||
NM_165586.5 | NP_724643.2 | 12 - 13 | 3880 | 59 | ||
NM_206056.5 | NP_995778.2 | 12 - 13 | 4166 | 59 | ||
GenBank mRNA | AF276964.1 | - | 11 - 12 | 3818 | 59 | |
AY069682.1 | - | 12 - 13 | 3835 | 59 | ||
BT100315.1 | - | 11 - 12 | 3845 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |