Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lig | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014503.2 | NP_001014503.1 | 6 - 7 | 2675 | 68 | |
NM_001169610.2 | NP_001163081.1 | 7 - 8 | 2668 | 68 | ||
NM_001169611.1 | NP_001163082.1 | 7 - 8 | 2692 | 68 | ||
NM_001169612.2 | NP_001163083.1 | 8 - 9 | 2734 | 68 | ||
NM_001259270.2 | NP_001246199.1 | 7 - 8 | 2668 | 68 | ||
NM_001259271.2 | NP_001246200.1 | 7 - 8 | 2668 | 68 | ||
NM_001273865.1 | NP_001260794.1 | 6 - 7 | 2675 | 68 | ||
NM_136504.5 | NP_610348.3 | 7 - 8 | 2668 | 68 | ||
NM_165586.5 | NP_724643.2 | 7 - 8 | 2737 | 68 | ||
NM_206056.5 | NP_995778.2 | 7 - 8 | 3023 | 68 | ||
GenBank mRNA | AF276964.1 | - | 6 - 7 | 2675 | 68 | |
AY069682.1 | - | 7 - 8 | 2692 | 68 | ||
BT100315.1 | - | 6 - 7 | 2641 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |