Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mtt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144147.2 | NP_001137619.1 | 14 - 15 | 4627 | 67 | |
NM_001273877.1 | NP_001260806.1 | 14 - 15 | 4627 | 67 | ||
NM_136542.3 | NP_610386.2 | 13 - 14 | 3667 | 67 | ||
NM_165609.3 | NP_724683.2 | 12 - 13 | 2721 | 67 | ||
NM_206058.2 | NP_995780.1 | 8 - 9 | 1193 | 67 | ||
GenBank mRNA | BT033068.1 | - | 8 - 9 | 1194 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor Drosophila melanogaster |