Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273901.1 | NP_001260830.1 | 4 - 5 | 1794 | 76 | |
NM_136667.3 | NP_610511.3 | 5 - 6 | 2066 | 76 | ||
NM_206066.3 | NP_995788.1 | 6 - 7 | 2538 | 76 | ||
GenBank mRNA | AJ289232.1 | - | 7 - 8 | 2408 | 76 | |
BT150239.1 | - | 6 - 7 | 2502 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |