Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144164.2 | NP_001137636.1 | 8 - 9 | 1197 | 96 | |
NM_001144166.3 | NP_001137638.2 | 4 - 5 | 6550 | 96 | ||
NM_001144168.2 | NP_001137640.1 | 6 - 7 | 720 | 96 | ||
NM_001259311.2 | NP_001246240.1 | 7 - 8 | 768 | 96 | ||
NM_001259312.2 | NP_001246241.1 | 6 - 7 | 6589 | 96 | ||
NM_001273931.1 | NP_001260860.1 | 4 - 5 | 6442 | 96 | ||
NM_001382042.1 | NP_001369074.1 | 4 - 5 | 6880 | 96 | ||
NM_001382043.1 | NP_001369075.1 | 6 - 7 | 6919 | 96 | ||
NM_001382045.1 | NP_001369077.1 | 4 - 5 | 6772 | 96 | ||
NM_136726.3 | NP_610570.1 | 5 - 6 | 559 | 96 | ||
NM_136727.4 | NP_610571.2 | 3 - 4 | 6343 | 96 | ||
NM_136728.4 | NP_610572.1 | 3 - 4 | 474 | 96 | ||
NM_165757.3 | NP_724910.1 | 9 - 10 | 1218 | 96 | ||
NM_165758.2 | NP_724911.1 | 8 - 9 | 1406 | 96 | ||
NM_165759.3 | NP_724912.1 | 3 - 4 | 1417 | 96 | ||
NM_206073.3 | NP_995795.1 | 4 - 5 | 598 | 96 | ||
GenBank mRNA | AY061565.1 | - | 3 - 4 | 441 | 96 | |
BT003217.1 | - | 9 - 10 | 1219 | 96 | ||
BT003435.1 | - | 8 - 9 | 1178 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |