Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp49a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259327.2 | NP_001246256.1 | 2 - 3 | 692 | 78 | |
NM_136744.4 | NP_610588.2 | 2 - 3 | 452 | 78 | ||
NM_206081.2 | NP_995803.1 | 2 - 3 | 398 | 78 | ||
GenBank mRNA | BT056244.1 | - | 2 - 3 | 444 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |