Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp49a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259327.2 | NP_001246256.1 | 4 - 5 | 1018 | 86 | |
NM_136744.4 | NP_610588.2 | 4 - 5 | 778 | 86 | ||
NM_206081.2 | NP_995803.1 | 4 - 5 | 724 | 86 | ||
GenBank mRNA | BT056244.1 | - | 4 - 5 | 770 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |