Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syx6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_206092.3 | NP_995814.1 | 3 - 4 | 1291 | 69 | |
NM_206093.3 | NP_995815.1 | 3 - 4 | 1040 | 69 | ||
NM_206094.3 | NP_995816.1 | 2 - 3 | 1396 | 69 | ||
GenBank mRNA | AY089525.1 | - | 2 - 3 | 1348 | 69 | |
BT001663.1 | - | 3 - 4 | 1289 | 69 | ||
BT003505.1 | - | 3 - 4 | 1038 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |