Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Obp50a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_166050.2 | NP_725385.1 | 1 - 2 | 184 | 72 | |
NM_206110.2 | NP_995832.1 | 1 - 2 | 184 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |