Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sema2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_058159.5 | NP_477507.1 | 3 - 4 | 635 | 67 | |
NM_134303.4 | NP_599130.1 | 3 - 4 | 536 | 67 | ||
NM_166178.3 | NP_725586.1 | 3 - 4 | 572 | 67 | ||
NM_206134.2 | NP_995856.2 | 3 - 4 | 495 | 67 | ||
GenBank mRNA | AY095026.1 | - | 3 - 4 | 565 | 67 | |
AY128437.1 | - | 3 - 4 | 480 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |