Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057775.4 | NP_477123.1 | 4 - 5 | 2118 | 70 | |
NM_166200.3 | NP_725623.2 | 3 - 4 | 2197 | 70 | ||
NM_206140.2 | NP_995862.1 | 4 - 5 | 2118 | 70 | ||
GenBank mRNA | BT031264.1 | - | 4 - 5 | 2085 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |