Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RhoGEF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259452.2 | NP_001246381.1 | 10 - 11 | 3726 | 85 | |
NM_001299586.1 | NP_001286515.1 | 10 - 11 | 3406 | 85 | ||
NM_057969.6 | NP_477317.1 | 10 - 11 | 3715 | 85 | ||
NM_206146.4 | NP_995868.1 | 10 - 11 | 3726 | 85 | ||
NM_206147.4 | NP_995869.1 | 9 - 10 | 2664 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |