Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: par-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014540.3 | NP_001014540.2 | 5 - 6 | 2338 | 77 | |
NM_001014542.3 | NP_001014542.1 | 5 - 6 | 2338 | 77 | ||
NM_001169737.2 | NP_001163208.1 | 5 - 6 | 2435 | 77 | ||
NM_001169738.2 | NP_001163209.1 | 5 - 6 | 2435 | 77 | ||
NM_001169739.2 | NP_001163210.1 | 5 - 6 | 2338 | 77 | ||
NM_001169740.2 | NP_001163211.1 | 5 - 6 | 2479 | 77 | ||
NM_206171.4 | NP_995893.1 | 5 - 6 | 2237 | 77 | ||
NM_206172.3 | NP_995894.2 | 5 - 6 | 2338 | 77 | ||
NM_206177.3 | NP_995899.2 | 5 - 6 | 2338 | 77 | ||
NM_206178.3 | NP_995900.1 | 5 - 6 | 2338 | 77 | ||
GenBank mRNA | AF258462.1 | - | 5 - 6 | 2313 | 77 | |
AF387635.1 | - | 5 - 6 | 2313 | 77 | ||
AF387636.1 | - | 4 - 5 | 2255 | 77 | ||
BT009940.1 | - | 5 - 6 | 2387 | 77 | ||
BT011020.1 | - | 5 - 6 | 2333 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |