Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bw | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299840.1 | NP_001286769.1 | 7 - 8 | 1646 | 62 | |
NM_079100.3 | NP_523824.1 | 7 - 8 | 1646 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |