Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Start1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259566.2 | NP_001246495.1 | 7 - 8 | 1100 | 73 | |
NM_138092.3 | NP_611936.2 | 7 - 8 | 933 | 73 | ||
NM_206217.3 | NP_995939.1 | 7 - 8 | 1100 | 73 | ||
GenBank mRNA | AY102683.1 | - | 7 - 8 | 1065 | 73 | |
AY455866.1 | - | 7 - 8 | 1082 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |