Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rno | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259599.2 | NP_001246528.1 | 7 - 8 | 2231 | 118 | |
NM_138163.2 | NP_612007.2 | 7 - 8 | 2179 | 118 | ||
NM_206222.3 | NP_995944.1 | 7 - 8 | 2395 | 118 | ||
GenBank mRNA | AY321364.1 | - | 7 - 8 | 2178 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Drosophila melanogaster |