Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: msn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274402.1 | NP_001261331.1 | 2 - 3 | 350 | 74 | |
NM_001274403.1 | NP_001261332.1 | 2 - 3 | 270 | 74 | ||
NM_001274404.1 | NP_001261333.1 | 2 - 3 | 407 | 74 | ||
NM_079940.5 | NP_524679.3 | 2 - 3 | 270 | 74 | ||
NM_206249.3 | NP_995971.2 | 2 - 3 | 270 | 74 | ||
NM_206250.3 | NP_995972.1 | 2 - 3 | 253 | 74 | ||
NM_206251.3 | NP_995973.1 | 2 - 3 | 264 | 74 | ||
NM_206252.3 | NP_995974.1 | 2 - 3 | 270 | 74 | ||
GenBank mRNA | BT031263.1 | - | 2 - 3 | 250 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |