Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kst | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104022.3 | NP_001097492.2 | 5 - 6 | 969 | 61 | |
NM_001202112.2 | NP_001189041.1 | 6 - 7 | 1031 | 61 | ||
NM_001274438.1 | NP_001261367.1 | 5 - 6 | 969 | 61 | ||
NM_001274439.1 | NP_001261368.1 | 6 - 7 | 997 | 61 | ||
NM_079176.3 | NP_523900.1 | 5 - 6 | 1466 | 61 | ||
NM_206265.3 | NP_995987.1 | 5 - 6 | 1466 | 61 | ||
NM_206266.3 | NP_995988.1 | 5 - 6 | 1466 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |