Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: enc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104026.2 | NP_001097496.1 | 5 - 6 | 1861 | 72 | |
NM_001104027.2 | NP_001097497.1 | 5 - 6 | 1804 | 72 | ||
NM_001274447.1 | NP_001261376.1 | 5 - 6 | 1819 | 72 | ||
NM_001274448.1 | NP_001261377.1 | 4 - 5 | 1361 | 72 | ||
NM_001274449.1 | NP_001261378.1 | 5 - 6 | 1804 | 72 | ||
NM_080026.3 | NP_524765.2 | 5 - 6 | 1804 | 72 | ||
NM_206270.2 | NP_995992.1 | 5 - 6 | 1819 | 72 | ||
GenBank mRNA | BT050449.1 | - | 5 - 6 | 1802 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |