Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dikar | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202125.2 | NP_001189054.1 | 4 - 5 | 1043 | 60 | |
NM_001274551.1 | NP_001261480.1 | 4 - 5 | 1043 | 60 | ||
NM_001274552.1 | NP_001261481.1 | 4 - 5 | 1043 | 60 | ||
NM_139798.4 | NP_648055.1 | 4 - 5 | 1043 | 60 | ||
NM_168179.3 | NP_729188.2 | 4 - 5 | 1043 | 60 | ||
NM_206285.1 | NP_996007.1 | 4 - 5 | 1006 | 60 | ||
GenBank mRNA | AY060386.1 | - | 4 - 5 | 1028 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |