Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: smid | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169900.2 | NP_001163371.1 | 8 - 9 | 2872 | 62 | |
NM_001274583.1 | NP_001261512.1 | 8 - 9 | 3018 | 62 | ||
NM_001274584.1 | NP_001261513.1 | 7 - 8 | 2824 | 62 | ||
NM_079235.4 | NP_523959.2 | 8 - 9 | 2875 | 62 | ||
NM_206287.3 | NP_996009.2 | 8 - 9 | 2785 | 62 | ||
GenBank mRNA | BT124778.1 | - | 8 - 9 | 3016 | 62 | |
X99207.1 | - | 2836 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |