Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tip60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272305.1 | NP_001259234.1 | 3 - 4 | 1511 | 78 | |
NM_131923.4 | NP_572151.1 | 3 - 4 | 2049 | 78 | ||
GenBank mRNA | AY051795.1 | - | 3 - 4 | 2004 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |