Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHOp24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001297933.1 | NP_001284862.1 | 2 - 3 | 571 | 79 | |
NM_131937.4 | NP_572165.1 | 2 - 3 | 571 | 79 | ||
GenBank mRNA | AY060633.1 | - | 2 - 3 | 530 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |